Learn More
METTLER TOLEDO pH Sensor InPro 2000i SG/450/9816
Supplier:  METTLER TOLEDO 52003525
Encompass
The InPro 2000i SG/450/9816 is an Intelligent Sensor Management (ISM) pH probe with an integrated temperature sensor for demanding applications. This 450 mm refillable pH sensor with Viscolyt electrolyte is a reliable pH measurement solution.
Catalog No. NC1697171