Learn More
Hach Company Hach pH Sensor, LCP Body, 5-Wire Convertible Mount, Low-Conductivity Glass Electrode, 10 ft Cable (6028P4)
Catalog No. NC3363328 Shop All Hach Company Products
Encompass
This LCP-bodied pH sensor is a convertible-mount electrode engineered for reliable pH measurement in low-conductivity solutions. It includes a built-in preamplifier and a 10 ft (3 m) cable for flexible installation in flow or immersion setups.
  • Uses differential electrode measurement technique for improved accuracy and reduced reference junction potential.
  • Replaceable salt bridge minimizes contamination and extends time between calibrations.
  • Built-in preamplifier allows long cable runs and robust signal transmission.
  • Convertible mounting with 1.5 in NPT threads for immersion or pipe-tee installation.
  • Designed for low-conductivity measurements using low-conductivity glass.
  • Operates from -5 to 95 °C and withstands flow rates up to 3 m/s.
Catalog No. NC3363328 Supplier Hach Company Supplier No. 6028P4
This LCP-bodied pH sensor is a convertible-mount electrode engineered for reliable pH measurement in low-conductivity solutions. It includes a built-in preamplifier and a 10 ft (3 m) cable for flexible installation in flow or immersion setups.
  • Uses differential electrode measurement technique for improved accuracy and reduced reference junction potential.
  • Replaceable salt bridge minimizes contamination and extends time between calibrations.
  • Built-in preamplifier allows long cable runs and robust signal transmission.
  • Convertible mounting with 1.5 in NPT threads for immersion or pipe-tee installation.
  • Designed for low-conductivity measurements using low-conductivity glass.
  • Operates from -5 to 95 °C and withstands flow rates up to 3 m/s.