Learn More
Sensorex pH Sensor,Ultem,DJ,BTD BNC
Catalog No. 501897520
Encompass_Preferred
The pH2000 has a double junction reference design to prevent reference fouling by heavy metals and other sample contaminants, low maintenance sealed gel reference design, durable Ultem body
Catalog No. 50-189-7520 Supplier Sensorex Supplier No. S200CD
The pH2000 has a double junction reference design to prevent reference fouling by heavy metals and other sample contaminants, low maintenance sealed gel reference design, durable Ultem body