Learn More
Sensorex pH Sensor, WTW,30K,DIN-M/4mmBP
Catalog No. 501897534
Encompass_Preferred
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and 30K automatic temperature compensation (ATC) and DIN M + 1 each 4 mm banana plug. For use with WTW.
Catalog No. 50-189-7534 Supplier Sensorex Supplier No. S295C
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and 30K automatic temperature compensation (ATC) and DIN M + 1 each 4 mm banana plug. For use with WTW.