Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ pH Soil Test Kit
SDPSCI_ED

Catalog No. s95000 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S95000 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95000 Supplier LaMotte™ Supplier No. 5024
Only null left
Add to Cart
Add to Cart

Includes

  • Reagents for 100 tests
  • Color chart
  • Plastic spot plate
  • Instructions

pH affects all mineral elements and the biological processes made available to plants from the soil.

  • Accurately test pH to determine lime requirements and ensure that mineral-rich soil is also fertile
  • pH range: 4.0 to 8.0
  • pH accuracy: ±0.5
  • pH resolution: 1

Specifications

For Use With (Application) Determine lime requirements and to insure that a mineral-rich soil is also a fertile one
Includes 20ml duplex indicator, 1 plastic spot plate, 1 duplex soil color chart, 1 lamotte™ soil handbook
Quantity 100 Tests
Class Size 1 Class
Product Type Soil pH Testing Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.