Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pH Standard, IUPAC pH 7.413, Hach Lange™

Catalog No. 501998094 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-8094 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-8094 Supplier Hach Company Supplier No. S11M005
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified buffer solution, traceable to standard reference materials (CRM). With a concentration of 7.413 ± 0.010 pH at 25°C, this pH buffer has a shelf life of 48 months from production date and is traceable to IUPAC pH scale and NIST.

  • Concentration of 7.413 ± 0.010 pH at 25°C
  • Shelf life of 48 months from production date
  • Traceable to IUPAC pH scale and NIST
  • Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
  • Certified buffer solution, traceable to standard reference materials (CRM)
  • Due to a special form of safekeeping the unopened standard is stable for several years
TRUSTED_SUSTAINABILITY

Specifications

Description Traceable to IUPAC pH scale and NIST
pH 7.413
Quantity 500 mL
Traceability to NIST Yes
ShelfLife 48 months from production date
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.