Learn More
Hach Company Certified Buffer Standard Solution, IUPAC, pH 9.180, 500 mL
Catalog No. 501998095 Shop All Hach Company Products
Encompass
pH Buffer IUPAC Standard Solution with Certificate of Conformity* and Traceability reporting exact pH buffer value with related expanded uncertainty (k=2). Traceable to IUPAC/NIST pH scale. Guaranteed shelf-life thanks to premium packaging. Colorless pH buffer solution. pH is 9.180 ±0.010 (k=2) at 25°C, 500 mL bottle.*Available for download from www.hach.com: Support/Service, Certificate of Analysis menu using product part number and lot number.
Catalog No. 50-199-8095 Supplier Hach Company Supplier No. S11M006US
pH Buffer IUPAC Standard Solution with Certificate of Conformity* and Traceability reporting exact pH buffer value with related expanded uncertainty (k=2). Traceable to IUPAC/NIST pH scale. Guaranteed shelf-life thanks to premium packaging. Colorless pH buffer solution. pH is 9.180 ±0.010 (k=2) at 25°C, 500 mL bottle.*Available for download from www.hach.com: Support/Service, Certificate of Analysis menu using product part number and lot number.