Learn More
Hach Company Buffer Solution, pH 10.01, 1 L
Supplier:  Hach Company LZW9472.99
Encompass
pH Buffer Standard Solution for pH electrode calibration, Colorless pH buffer, pH is 10.01 ±0.02 at 25°C, 1 L bottle
Catalog No. 50-199-8013