Learn More
Hach Company Buffer Solution, pH 10.01, 250 mL
Supplier: Hach Company LZW9471.99
pH Buffer Standard Solution for pH electrode calibration, Colorless pH buffer, pH is 10.01 ±0.02 at 25°C, 250 mL bottle