Learn More
Hach Company Buffer Solution, pH 4.01, 250 mL
Supplier:  Hach Company LZW9463.99
Encompass
pH Buffer Standard Solution for pH electrode calibration, Colorless pH buffer, pH is 4.01 ±0.02 at 25°C, 250 mL bottle
Catalog No. 50-199-8007