Learn More
Hach Company Buffer Solution, pH 7.00, 250mL
Supplier:  Hach Company LZW9464.97
Encompass
pH Buffer Standard Solution for pH electrode calibration, Colorless pH buffer, pH is 7.00 ±0.02 at 25°C, 250 mL bottle
Catalog No. 50-199-8008