Learn More
Hach Company pH Electrode Storage Solution Powder Pillows, pk/20
Supplier: Hach Company 2657364
pH Electrode Storage Solution Powder Pillows. Each pillow makes 25 mL of storage solution. Pack of 20 Powder Pillows