Learn More
Hach Company pH Electrode Storage Solution, 50 mL
Catalog No. 501996629 Shop All Hach Company Products
Encompass
Potassium Chloride (3M) in 50 mL pour-top bottle.
Catalog No. 50-199-6629 Supplier Hach Company Supplier No. 2756526
Potassium Chloride (3M) in 50 mL pour-top bottle.