Learn More
Cole-Parmer Oakton All-in-One pH/ATC Electrode, Epoxy, Sealed, Double-Junction, 12 mm OD; 3 ft Sub-Mini/BNC
Catalog No. NC9395076 Shop All Cole-Parmer Products
Encompass_Preferred
All-in-one combination, pH/Temp electrode, 3-ft cable, epoxy body, double-junction, gel filled, sub-miniature audio ATC, BNC, 12 mm dia.
Catalog No. NC9395076 Supplier Cole-Parmer Supplier No. 3581172
All-in-one combination, pH/Temp electrode, 3-ft cable, epoxy body, double-junction, gel filled, sub-miniature audio ATC, BNC, 12 mm dia.