Learn More
Cole-Parmer Oakton Not for individual sale. Use 35811-71 instead to re-order this electrode.
Catalog No. NC0980666 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. NC0980666 Supplier Cole-Parmer Supplier No. 3581198
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.