Learn More
Hach Company pH Paper, 0 - 14 pH Range, 100/pk
Supplier: Hach Company 2601300
Provides a simple, fast, economical method of pH measurement. Obtain accurate results in weakly buffered, turbid and lightly colored samples. Measures 0-14 pH.