Learn More
Fisherbrand™ pH Test Strips
Catalog No. 13640521 Shop All Fisherbrand Products
Change view
Click to view available options
pH Range:
0 to 14
0 to 6
2 to 9
4 to 7
4.5 to 10
6 to 10
7 to 14
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
13-640-521 0 to 14
13-640-522 0 to 6
13-640-527 2 to 9
13-640-523 4 to 7
13-640-524 4.5 to 10
13-640-525 6 to 10
13-640-526 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-640-521
Add to Cart
Add to Cart
Conveniently packed in flip-top vials, these multipad pH strips provide excellent color separation over the entire pH scale with clear multicolor matches at every interval.
  • Non bleed, multiple pad color match
  • Dip and read instant color match
  • Flip-top vial stays closed when dropped, but is easy to open with one-hand
  • Sturdy vial keeps strips safe from the elements
  • 100 strips per vial
Specifications
pH Range 0 to 14
No. of Tests 100
Range Narrow