Learn More
Grainger pH Test Strips 6 in L 0 to 13 pH PK10
Catalog No. 502753531
Encompass_Preferred
The Hydrion pH Test Strips 1.0 pH measure pH levels between 1.0 and 3.0 with 0.5 pH increments. They provide accurate pH testing for acids and are constructed of chemically inert and stain resistant materials. The test strips come in a plastic dispenser vial containing 100 strips. To test, dip the strip into solution for 1 second and compare color to chart on vial label. These strips are useful for testing pH in the food industry and laboratories. They provide fast, convenient, and accurate pH testing.
Catalog No. 50-275-3531 Supplier Grainger Supplier No. 14G847
The Hydrion pH Test Strips 1.0 pH measure pH levels between 1.0 and 3.0 with 0.5 pH increments. They provide accurate pH testing for acids and are constructed of chemically inert and stain resistant materials. The test strips come in a plastic dispenser vial containing 100 strips. To test, dip the strip into solution for 1 second and compare color to chart on vial label. These strips are useful for testing pH in the food industry and laboratories. They provide fast, convenient, and accurate pH testing.