Learn More
Hach Company Titration Application Pack Food & Beverage: pH & Total Acidity (Milk, Tomato Sauce, Fruit Juice)
Catalog No. 501997513 Shop All Hach Company Products
Encompass
Application Pack for the determination of pH and Total Acidity in food & beverages. Suited for Titralab AT1000 Series, Automatic Titrator Models AT1102/AT1112/AT1122/AT1222.
Catalog No. 50-199-7513 Supplier Hach Company Supplier No. AP0008.AT1102
Application Pack for the determination of pH and Total Acidity in food & beverages. Suited for Titralab AT1000 Series, Automatic Titrator Models AT1102/AT1112/AT1122/AT1222.