Learn More
Inorganic Ventures pH 7 YELLOW CALIBRATION STD 4
SDP
Catalog No. NC0834431
Encompass_Preferred
Inorganic Ventures' pH 7 Yellow Standard is a 4 liter certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration .
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC0834431 Supplier Inorganic Ventures Supplier No. PHYELLOW74L
Inorganic Ventures' pH 7 Yellow Standard is a 4 liter certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration .
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search