Learn More
HANNA INSTRUMENTS INC pH 10.01 Calibration Buffer Sachets (25 x 20 mL)
SDP
Catalog No. NC1811110
Encompass
HI70010P is a premium quality pH 10.01 calibration buffer solution. Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened. Hanna's line of calibration buffers have been specially formulated to have an expiration of 2 years from the date of manufacture for an unopened sachet. The HI70010P is for 25 sachets containing 20 mL of buffer.Color coded label for easy pH buffer value identificationAccuracy of +/- 0.01 pH @ 25°CTemperature chart of the actual pH value at various temperatures printed on each sachet
Catalog No. NC1811110 Supplier HANNA INSTRUMENTS INC Supplier No. HI70010P
HI70010P is a premium quality pH 10.01 calibration buffer solution. Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened. Hanna's line of calibration buffers have been specially formulated to have an expiration of 2 years from the date of manufacture for an unopened sachet. The HI70010P is for 25 sachets containing 20 mL of buffer.Color coded label for easy pH buffer value identificationAccuracy of +/- 0.01 pH @ 25°CTemperature chart of the actual pH value at various temperatures printed on each sachet