Learn More
Cole-Parmer Oakton Traceable pH Standard Buffer with Calibration, Clear, pH 10; 500 mL
Catalog No. NC0036230 Shop All Cole-Parmer Products
Encompass_Preferred
Oakton Traceable pH Standard Buffer with Calibration, Clear, pH 10; 500 mLNIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratoryChoose from clear or color-coded standards from common calibration points500 mL (16 fl.oz.) capacityCalibrate at pH 10
Catalog No. NC0036230 Supplier Cole-Parmer Supplier No. 0065104
Oakton Traceable pH Standard Buffer with Calibration, Clear, pH 10; 500 mLNIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratoryChoose from clear or color-coded standards from common calibration points500 mL (16 fl.oz.) capacityCalibrate at pH 10