Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PH111 pH/mV Bench Meter Standard Kit
SCI_ED

Catalog No. S81897 Shop All Thermo Scientific Products
Change view
Click to view available options
Range (pH):
-2 to 18
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
S81897 -2 to 18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81897 Supplier Thermo Scientific™ Supplier No. LSTAR1115/EMD

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Orion Lab Star PH111 pH bench meters measure pH or ORP with mV and temperature, feature a large backlit display for easy viewability, and offer small footprint to save bench space. Ideal for simplified, budget-friendly lab testing and data recording.

Orion Lab Star PH111 pH Meter Features

  • Budget-friendly pH meter for simplified pH and ORP testing
  • Display shows pH, mV and temperature simultaneously for accurate data recording
  • Measure -2 to 18 pH, -2000 to +2000 mV and -5 to 105 °C or 23 to 221 °F to ensure comprehensive measurement capabilities
  • Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance
  • Large 5 inch backlit display makes viewing and recording measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Onscreen stabilizing and ready indicators help identify when readings are complete
  • pH electrode condition icon helps ensure reliable electrode performance
  • Date and time data stamped data and calibration logs support GLP compliance
  • View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements
  • Use 500-point data log with date and time stamp to record key measurements
  • Output data and calibration logs to a printer or computer for long-term data storage
  • Interchangeable wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets

Meter and Meter Kits

  • LSTAR1110 is the pH meter and stand option, which allows you to purchase your pH electrode separately
  • LSTAR1115 is the standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use
  • LSTAR116 is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore
  • LSTAR117 is the low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate
  • LSTAR1118 is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges
TRUSTED_SUSTAINABILITY

Specifications

Description pH/mV Bench Meter Standard Kit
Accuracy (Temperature) ±0.3°C (±0.5°F)
Accuracy (mV) ±0.2% or ±0.05% (Whichever is Greater)
Data Management Details Transfer Single or All Data Points to Printer or Computer
Outputs Computer or Printer (STARA-106)
Accuracy (pH) ±0.005, ±1 LSD
Includes Refillable Epoxy Body pH/ATC Electrode, pH 4/7/10 Buffer, Storage and Cleaning Solution, 60 mL Kit Meter with Electrode Stand, PC Cable, Power Adapter
For Use With (Application) Drinking Water, Tap Water, Buffers (Non-TRIS) and General Purpose Student Use
Measurement Modes pH with mV and Temperature, ORP with mV and Temperature
Calibration pH, ORP, Temperature
Range (pH) -2 to 18
Resolution (pH) 0.001, 0.01, 0.1
Calibration Points (pH) 1 to 5 Points
Range (mV) ±2000 mV with ±250 mV Offset
Resolution (mV) 0.1 mV
Calibration Points 1 (ORP)
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 23°F to 221°F
Resolution (Temperature) 0.1°C (0.1°F)
Temperature Selection Manual, Automatic
Temperature Probe Calibration 1 Point Offset Calibration
Display Type LCD, Backlit
Backlight Option Yes
Memory Non-volatile
Data Points 500 with Date and Time Stamp
Log Function Type Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode
Alarm Output Calibration Reminder Alarm
Inputs BNC for pH or ORP, 8-pin MiniDIN for ATC
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
Dimensions (L x W x H) 198.3 x 155.2 x 61.7 mm (7.81 x 6.11 x 2.43 in.)
Weight (Metric) 700 g
Weight (English) 1.54 lb.
Warranty 3 Years (Meter)
Power Supply Universal
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.