Learn More
Cole-Parmer pH 4 buffer, red, 4L, NIST-traceable
Supplier:  Cole-Parmer 0065176
Encompass_Preferred
Oakton Traceable pH Standard Buffer with Calibration, Red, pH 4; 4 L, NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratoryChoose from clear or color-coded standards from common calibration points4 L (128 fl.oz.) capacityCalibrate at pH 4Easily identify red color-coded formula
Catalog No. NC1504364