Learn More
Hach Company GLI pH 7.0 Buffer, Std Cell 3.5 L
Catalog No. 501997401 Shop All Hach Company Products
Encompass
Standard Cell Solution for Hach pHD Differential pH and ORP Sensors, packaged in resealable 500 mL bottle. Used to replenish standard cell chamber while replacing salt bridge.
Catalog No. 50-199-7401 Supplier Hach Company Supplier No. 25M1A1001123
Standard Cell Solution for Hach pHD Differential pH and ORP Sensors, packaged in resealable 500 mL bottle. Used to replenish standard cell chamber while replacing salt bridge.