Learn More
Hach Company GLI pH 7.0 Buffer, Std Cell, 500 mL
Catalog No. 501997400 Shop All Hach Company Products
Encompass
Standard Cell Solution for Hach pHD Differential pH and ORP Sensors, packaged in resealable 500 mL bottle. Used to replenish standard cell chamber while replacing salt bridge.
Catalog No. 50-199-7400 Supplier Hach Company Supplier No. 25M1A1001115
Standard Cell Solution for Hach pHD Differential pH and ORP Sensors, packaged in resealable 500 mL bottle. Used to replenish standard cell chamber while replacing salt bridge.