Learn More
MSC Betco pH7Q Disinfectant, Neutral, Lemon Scent, 1 gal, 4/Pk
Catalog No. 502884669
Encompass
Betco pH7Q Neutral Disinfectant is a versatile germicidal detergent and deodorant that cleans, disinfects, and deodorizes in a single step. Its neutral pH formula is safe for damp mopping highly polished floors without dulling or leaving a film. It effectively kills the SARS-CoV-2 virus on hard non-porous surfaces within one minute.
  • Effective against antibiotic-resistant strains, VRE, and MRSA
  • Neutral pH formulation that will not dull, haze, or attack floor finish
  • Leaves no residue or film on surfaces
  • Kills SARS-CoV-2 virus on hard non-porous surfaces in one minute
  • Pleasant lemon fragrance for a fresh scent
  • Liquid form for easy application
  • Contains quaternary ammonium compounds as active ingredients
  • Recommended dilution of 2 oz per gallon for effective use
Catalog No. 50-288-4669 Supplier MSC Supplier No. 72529316
Betco pH7Q Neutral Disinfectant is a versatile germicidal detergent and deodorant that cleans, disinfects, and deodorizes in a single step. Its neutral pH formula is safe for damp mopping highly polished floors without dulling or leaving a film. It effectively kills the SARS-CoV-2 virus on hard non-porous surfaces within one minute.
  • Effective against antibiotic-resistant strains, VRE, and MRSA
  • Neutral pH formulation that will not dull, haze, or attack floor finish
  • Leaves no residue or film on surfaces
  • Kills SARS-CoV-2 virus on hard non-porous surfaces in one minute
  • Pleasant lemon fragrance for a fresh scent
  • Liquid form for easy application
  • Contains quaternary ammonium compounds as active ingredients
  • Recommended dilution of 2 oz per gallon for effective use