Learn More
Medchemexpress LLC PHA-408 | 503555-55-3 | MFCD17166977 | 99.4% | 560.02 g·mol⁻¹ | C29H27ClFN7O2 | 5 MG
SDP
Catalog No. 5000221260
Encompass_Preferred
PHA-408 is a potent, selective, orally active inhibitor of IκB kinase-2 (IKK-2/IKKβ) used in preclinical research to investigate inflammatory signaling and lung inflammation models. It is provided as a high-purity solid suitable for laboratory assays and biochemical studies.
  • Potent and selective inhibitor of IκB kinase-2 for signaling studies.
  • Demonstrated anti-inflammatory activity in LPS- and cigarette smoke-mediated lung inflammation models.
  • Supplied as a high-purity solid (≈99.4%).
  • Chemical formula C29H27ClFN7O2; molecular weight 560.02 g·mol⁻¹.
  • Stable when stored at recommended low temperatures for long-term use.
Catalog No. 50-002-21260 Supplier Medchemexpress LLC Supplier No. HY141805MG
PHA-408 is a potent, selective, orally active inhibitor of IκB kinase-2 (IKK-2/IKKβ) used in preclinical research to investigate inflammatory signaling and lung inflammation models. It is provided as a high-purity solid suitable for laboratory assays and biochemical studies.
  • Potent and selective inhibitor of IκB kinase-2 for signaling studies.
  • Demonstrated anti-inflammatory activity in LPS- and cigarette smoke-mediated lung inflammation models.
  • Supplied as a high-purity solid (≈99.4%).
  • Chemical formula C29H27ClFN7O2; molecular weight 560.02 g·mol⁻¹.
  • Stable when stored at recommended low temperatures for long-term use.