Learn More
TARGETMOL CHEMICALS INC PHA-665752 25MG
SDP
Catalog No. 502579534
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PHA-665752 is an effective specific and ATP-competitive c-Met inhibitor (IC50 9 nM) >50-fold selectivity for c-Met than STKs or RTKs. purity: 99%
Catalog No. 50-257-9534 Supplier TARGETMOL CHEMICALS INC Supplier No. T612825MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PHA-665752 is an effective specific and ATP-competitive c-Met inhibitor (IC50 9 nM) >50-fold selectivity for c-Met than STKs or RTKs. purity: 99%