Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pha-665752 | 477575-56-7 | MFCD18632554; MFCD07772270 | 99.9% | 641.61 g/mol | C32H34Cl2N4O4S | 5 MG
SDP

Catalog No. 5000263292
Encompass_Preferred

PHA-665752 is a selective, ATP-competitive inhibitor of the c-Met (HGFR) receptor tyrosine kinase used in preclinical research to induce apoptosis, cause cell-cycle arrest, and reduce tumor growth in vitro and in vivo. It demonstrates high potency and selectivity versus other kinases and is supplied for research use only.

  • Selective ATP-competitive c-Met inhibitor with Ki ≈ 4 nM and IC50 ≈ 9 nM.
  • High purity solid (approximately 99.9%) suitable for biochemical assays.
  • Molecular formula C32H34Cl2N4O4S; molecular weight 641.61 g/mol.
  • Storage: powder at -20°C for long term; in solution store at -80°C when possible.
  • Supplied in small preweighed milligram quantities for laboratory research.

Catalog No. 50-002-63292 Supplier Medchemexpress LLC Supplier No. HY111075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PHA-665752 is a selective, ATP-competitive inhibitor of the c-Met (HGFR) receptor tyrosine kinase used in preclinical research to induce apoptosis, cause cell-cycle arrest, and reduce tumor growth in vitro and in vivo. It demonstrates high potency and selectivity versus other kinases and is supplied for research use only.

  • Selective ATP-competitive c-Met inhibitor with Ki ≈ 4 nM and IC50 ≈ 9 nM.
  • High purity solid (approximately 99.9%) suitable for biochemical assays.
  • Molecular formula C32H34Cl2N4O4S; molecular weight 641.61 g/mol.
  • Storage: powder at -20°C for long term; in solution store at -80°C when possible.
  • Supplied in small preweighed milligram quantities for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.