Learn More
Medchemexpress LLC PHA-767491 | 845714-00-3 | MFCD09839931 | 100.0% | 213.24 g·mol^-1 | C12H11N3O | 5 MG
SDP
Catalog No. 5000223358
Encompass_Preferred
PHA-767491 is a small-molecule dual inhibitor of Cdc7 and Cdk9 kinases for laboratory research. It shows low-nanomolar inhibition of Cdc7 (IC50 10 nM) and Cdk9 (IC50 34 nM) and is supplied with manufacturer documentation and batch-specific quality data.
  • Inhibits Cdc7 and Cdk9 with low-nanomolar potency.
  • Suitable for cell-cycle and transcriptional research.
  • High HPLC purity reported for tested batches.
  • Supplied with a certificate of analysis and technical datasheet.
  • Available in small mass sizes for preparative and screening use.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-23358 Supplier Medchemexpress LLC Supplier No. HY134615MG
PHA-767491 is a small-molecule dual inhibitor of Cdc7 and Cdk9 kinases for laboratory research. It shows low-nanomolar inhibition of Cdc7 (IC50 10 nM) and Cdk9 (IC50 34 nM) and is supplied with manufacturer documentation and batch-specific quality data.
  • Inhibits Cdc7 and Cdk9 with low-nanomolar potency.
  • Suitable for cell-cycle and transcriptional research.
  • High HPLC purity reported for tested batches.
  • Supplied with a certificate of analysis and technical datasheet.
  • Available in small mass sizes for preparative and screening use.
  • For research use only; not for human or veterinary use.