Learn More
Medchemexpress LLC PHA-767491 hydrochloride | 942425-68-5 | MFCD11519962 | 99.5% | 249.70 | C12H12ClN3O | 5 MG
SDP
Catalog No. 5000222159
Encompass_Preferred
PHA-767491 hydrochloride is a small-molecule research inhibitor that selectively targets Cdc7 and Cdk9 kinases. It is used in biochemical and cellular studies to probe DNA replication initiation, transcriptional regulation, and cancer cell proliferation. The compound exhibits low-nanomolar biochemical potency and measurable cellular activity, and is supplied as a high-purity solid for laboratory research.
  • Potent dual Cdc7/Cdk9 inhibition with low-nanomolar IC50 values.
  • Demonstrated cellular antiproliferative activity in multiple cancer cell lines.
  • High purity suitable for biochemical and cellular assays.
  • Solid, light yellow to yellow appearance for convenient handling.
  • Stable under recommended storage conditions for research use.
  • Analytical documentation available, including datasheet, COA, and SDS.
Catalog No. 50-002-22159 Supplier Medchemexpress LLC Supplier No. HY13461A5MG
PHA-767491 hydrochloride is a small-molecule research inhibitor that selectively targets Cdc7 and Cdk9 kinases. It is used in biochemical and cellular studies to probe DNA replication initiation, transcriptional regulation, and cancer cell proliferation. The compound exhibits low-nanomolar biochemical potency and measurable cellular activity, and is supplied as a high-purity solid for laboratory research.
  • Potent dual Cdc7/Cdk9 inhibition with low-nanomolar IC50 values.
  • Demonstrated cellular antiproliferative activity in multiple cancer cell lines.
  • High purity suitable for biochemical and cellular assays.
  • Solid, light yellow to yellow appearance for convenient handling.
  • Stable under recommended storage conditions for research use.
  • Analytical documentation available, including datasheet, COA, and SDS.