Learn More
Cayman Chemical PHA-767491 (hydrochloride), Purity: ≥98%, CAS: 942425-68-5, 5 mg
SDP
Catalog No. 502965174
Encompass_Preferred
An inhibitor of Cdc7 kinase (IC50 = 10 nM) as well as Cdk9, (IC50 = 34 nM); induces apoptotic cell death in multiple cancer cell types at 10 µM
Catalog No. 50-296-5174 Supplier Cayman Chemical Supplier No. 182185 MG
An inhibitor of Cdc7 kinase (IC50 = 10 nM) as well as Cdk9, (IC50 = 34 nM); induces apoptotic cell death in multiple cancer cell types at 10 µM