Learn More
Selleck Chemical LLC PHA-793887 50mg 718630-59-2
SDP
Catalog No. 508487
Encompass_Preferred
PHA-793887 is a novel and potent inhibitor of CDK2, CDK5 and CDK7 with IC50 of 8 nM, 5 nM and 10 nM. It is greater than 6-fold more selective for CDK2, 5, and 7 than CDK1, 4, and 9. PHA-793887 induces cell-cycle arrest and apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-848-7 Supplier Selleck Chemical LLC Supplier No. S148750MG
PHA-793887 is a novel and potent inhibitor of CDK2, CDK5 and CDK7 with IC50 of 8 nM, 5 nM and 10 nM. It is greater than 6-fold more selective for CDK2, 5, and 7 than CDK1, 4, and 9. PHA-793887 induces cell-cycle arrest and apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.