Learn More
Vector Laboratories Phaseolus Vulgaris Erythroagglutinin (PHA-E), Unconjugated 5mg
SDP
Catalog No. 502045670
Encompass
Phaseolus vulgaris agglutinin is the name ascribed to a family of lectins, each of which consists of four subunits. There are two different types of subunits. One appears to be involved primarily in red cell agglutination and has been designated the “E” subunit (for erythroagglutinin). The other type is involved in lymphocyte agglutination and mitogenic activity and has been termed the “L” subunit (for leucoagglutinin). These subunits combine to produce five isolectins. 
Catalog No. 50-204-5670 Supplier Vector Laboratories Supplier No. L11205
Add to Cart
Add to Cart
Phaseolus vulgaris agglutinin is the name ascribed to a family of lectins, each of which consists of four subunits. There are two different types of subunits. One appears to be involved primarily in red cell agglutination and has been designated the “E” subunit (for erythroagglutinin). The other type is involved in lymphocyte agglutination and mitogenic activity and has been termed the “L” subunit (for leucoagglutinin). These subunits combine to produce five isolectins. 