Learn More
Phaclofen, Tocris Bioscience™

Description
Specifications
Specifications
| CAS | 114012-12-3 |
| Quantity | 50 mg |
| Molecular Formula | C9H13ClNO3P |
| Synonym | phaclofen, 3-amino-2-4-chlorophenyl propyl phosphonic acid, phosphonic acid, 3-amino-2-4-chlorophenyl propyl, phosphonic acid,p-3-amino-2-4-chlorophenyl propyl, 3-amino-2-4-chlorophenyl propylphosphonic acid, phaclofen, solid, beta-4-chlorophenyl-3-aminopropylphosphonic acid, acmc-20djlx, biomol-nt_000256 |
| InChI Key | VSGNGLJPOGUDON-UHFFFAOYSA-N |
| SMILES | C1=CC(=CC=C1C(CN)CP(=O)(O)O)Cl |
| IUPAC Name | [3-amino-2-(4-chlorophenyl)propyl]phosphonic acid |
| Molecular Weight (g/mol) | 249.631 |
| PubChem CID | 1641 |
| Formula Weight | 249.63 |
| Show More |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.