Learn More
Aviva Systems Biology Corp PHACS Antibody - middle region (ARP48970_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48970_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PHACS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PHACS. Alternate names: ACS, PHACSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59978