Learn More
Aviva Systems Biology Corp PHACTR1 Antibody - middle region (ARP55397_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55397_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHACTR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PHACTR1. Alternate names: KIAA1733, MGC126575, MGC126577, RPEL, RPEL1, dJ257A7.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53752