Learn More
Aviva Systems Biology Corp PHACTR3 Antibody - C-terminal region (ARP53447_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53447_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHACTR3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PHACTR3. Alternate names: C20orf101, H17739, MGC117178, SCAPIN1, SCAPININApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53716