Learn More
Aviva Systems Biology Corp Phactr3 Antibody - middle region (ARP53446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Phactr3. It was validated on Western Blot. . Alternate names: 1500003N10Rik, 4930415A02Rik, H17739, KIAA4224, SCAPIN1, mKIAA4224, scapinin, Scapin1, ScapininApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-840