Learn More
PHADEBAS INC PHADEBAS AMYLASE 50TABLETS
Catalog No. NC9444648
Encompass
Phadebas Amylase Test; Used for both qualitative and quantitative assessment of the Amylase enzyme; Consists of starch tablets in which a blue dye has been chemically bound to the starch
Catalog No. NC9444648 Supplier PHADEBAS INC Supplier No. 1301
Phadebas Amylase Test; Used for both qualitative and quantitative assessment of the Amylase enzyme; Consists of starch tablets in which a blue dye has been chemically bound to the starch