Learn More
PHADEBAS INC Phadebas Amylase test, 50 tablets
Catalog No. NC0838773
Encompass
Phadebas Amylase test tablets are used for accurately determining alpha-amylase activity for a wide variety of applications, from different fields in the food industry to biotech and detergents.
Catalog No. NC0838773 Supplier PHADEBAS INC Supplier No. 12865983
Phadebas Amylase test tablets are used for accurately determining alpha-amylase activity for a wide variety of applications, from different fields in the food industry to biotech and detergents.