Learn More
PHADEBAS INC Phadebas Forensic A3S, 25 sheets, pre-coated paper, 395 mm length, 300 mm width, press test, alpha-amylase detection, blue color reaction, low animal amylase interference
Catalog No. NC9621130
Encompass
Phadebas Forensic A3S, 25 sheets, pre-coated paper, 395 mm length, 300 mm width, press test, alpha-amylase detection, blue color reaction, low animal amylase interference
Catalog No. NC9621130 Supplier PHADEBAS INC Supplier No. 1315
Phadebas Forensic A3S, 25 sheets, pre-coated paper, 395 mm length, 300 mm width, press test, alpha-amylase detection, blue color reaction, low animal amylase interference