Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phadia™ 1000 Immunoassay Analyzers
GSA_VA

Catalog No. 12380001p
Change view
Click to view available options
Throughput:
1500 results per 8 hour shift
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Throughput
12-380-001P 1500 results per 8 hour shift
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-380-001P Supplier Thermo Scientific™ Supplier No. 12380001
Only null left
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Provide an optimal allergy testing solution using advanced, state-of-the-art technology with the Phadia™ 1000 Laboratory System.

Provide optimal allergy testing solutions using advanced, state-of-the-art technology. The Phadia™ 1000 Laboratory System is the optimal solution for the large, cost-conscious laboratory to meet the growing need for allergy testing. Phadia 1000 is designed for high-throughput laboratories, delivering 2,000 to 10,000 results per week, with proven performance in laboratories worldwide.

Delivering the first result in 100 minutes and then one result every 15 seconds, Phadia 1000 is designed for the high-volume laboratory delivering approximately 2,000 to 10,000 tests per week.

Phadia 1000 can be connected to a Laboratory Automation System for fast and reliable transportation of samples. All instruments of the Phadia Laboratory Systems family can be clustered, allowing laboratories to share resources, and thereby ensure that samples are processed as efficiently as possible.

High Productivity and Automations

  • Throughput: >1200 results in one assay
  • Fully Automated processing of ImmunoCAP assays
  • Continuous Random Access
  • Onboard reagent storage, up to 10,000 tests and all reagents
  • Positive identification and full traceability of all samples and reagents
  • Accurate, standardized and reproducible test results
  • True quantitative measurements

Cost Effectiveness

  • Large panel of standardized, high-quality allergens, allergen components and antigens available
  • 40μl serum or plasma needed per test, plus dead volume depending on the sample tube
  • 28-day storage of calibration curves

Remote diagnostics
The ImmunoCAP Specific IgE blood test assay portfolio covers a wide range of tests including: grass, weed and tree pollens, microorganisms, mites, cats, dogs and other furred animals, insects, venoms and food allergens.

Throughput and Productivity
Phadia systems are all highly automated, requiring minimal hands-on time. Choosing the system with the capacity matching your needs for throughput ensures optimal productivity.

Specialized Software
Phadia Information Data Manager (IDM) Software

  • Allows 28 days storage of standard curves
  • Allows export of results in several formats for connectivity with Laboratory Information Systems (LIS)
  • Offers flexible report options
  • Offers multilingual support

Remote Diagnostics
Phadia LabCommunity provides customers with enhanced support and troubleshooting assistance that reduces system downtime. Features include:

  • Remote collection of data needed for service and support activities
  • Proactive system health status check
  • Automatic reporting of Quality Club results to Phadia

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Phadia 1000 Immunoassay Analyzer
Throughput 1500 results per 8 hour shift
Ambient Temperature 18°C to 32°C
Depth (English) 40 in.
Height (English) 65 in.
Width (English) 71 in.
Voltage 230 V
Mainframe Connection Protocol ASTM, MasterCAP™, Phamas™
Max. Networked Workstations 5 systems of each type
Operating System Windows™2000/ Windows XP
Weight (English) 1700 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.