Learn More
Thermo Scientific™ Phadia™ β 2-Glycoprotein I Antibodies for Antiphospholipid Syndrome
Catalog No. 14553301P Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
EliA Beta2-Glycoprotein I IgA
EliA Beta2-Glycoprotein I IgG
EliA Beta2-Glycoprotein I IgM
Quantity:
2 x 12 Tests
4 x 12 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
14-553-301P EliA Beta2-Glycoprotein I IgM 4 x 12 Tests
14-553-201P EliA Beta2-Glycoprotein I IgG 4 x 12 Tests
14-553-101P EliA Beta2-Glycoprotein I IgA 2 x 12 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-553-301P Supplier Thermo Scientific™ Supplier No. 14553301
Add to Cart
Add to Cart
This item is not returnable. View return policy
Intended for the in vitro quantitative measurement of antibodies directed to β2-Glycoprotein I in human serum and plasma to aid in the diagnosis of antiphospholipid syndrome (APS) and to evaluate the thrombotic risk in patients with systemic lupus erythematosus (SLE).
If present in the patient's specimen, antibodies to β2-Glycoprotein I bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG (IgM, IgA) antibodies (EliA IgG, IgM, IgA Conjugate™) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA 2-Glycoprotein I IgG (IgM, IgA) Wells are coated with human β2-Glycoprotein I antigen
  • The higher the responseβ value, the more specific IgG (IgM, IgA) is present in the specimen
  • All conjugates are isotype-specific -- mixed conjugate is not available
TRUSTED_SUSTAINABILITY
Specifications
Type EliA Beta2-Glycoprotein I IgM
For Use With (Application) In-vitro quantitative measurement of Antibodies directed to beta2-Glycoprotein I in Human Serum and Plasma to aid in the diagnosis of antiphospholipid syndrome
Quantity 4 x 12 Tests