Learn More
Thermo Scientific™ Phadia™ 2500 Immunoassay Analyzers
Description
Provide optimal allergy and autoimmunity testing solutions using advanced, state-of-the-art technology. The Phadia™ 2500 Laboratory System is the optimal solution for the large, cost-conscious laboratory to meet the growing need for allergy testing. Phadia 2500 manages very high throughputs, delivering 10,000 to 25,000 results per week, with proven performance in laboratories worldwide.
Phadia 2500 delivers the first result within 100 minutes from the start time and then processes 60 tests per hour. The instrument is ideal for labs performing approximately 10,000 to 25,000 tests per week.
Phadia 2500 can be connected to a Laboratory Automation System for fast and reliable transportation of samples. All instruments of the Phadia Laboratory Systems family can be clustered, allowing laboratories to share resources, and thereby ensure that samples are processed as efficiently as possible.
High Productivity and Automations
- Throughput: 3840 results in one assay
- Automated processing of ImmunoCAP
- Continuous Random Access
- Onboard reagent storage
- Accurate, standardized and reproducible test results
- True quantitative measurements
Cost Effectiveness
- Large panel of standardized, high-quality allergens, allergen components and antigens available
- 40μl serum or plasma needed per test, plus dead volume depending on the sample tube
- 28-day storage of calibration curves
Remote diagnostics
The ImmunoCAP Specific IgE blood test assay portfolio covers a wide range of tests including: grass, weed and tree pollens, microorganisms, mites, cats, dogs and other furred animals, insects, venoms and food allergens.
Throughput and Productivity
Phadia systems are all highly automated, requiring minimal hands-on time. Choosing the system with the capacity matching your needs for throughput ensures optimal productivity.
Specialized Software
Phadia Information Data Manager (IDM) Software
- Allows 28 days storage of standard curves
- Allows export of results in several formats for connectivity with Laboratory Information Systems (LIS)
- Offers flexible report options
- Offers multilingual support
Remote Diagnostics
Phadia LabCommunity provides customers with enhanced support and troubleshooting assistance that reduces system downtime. Features include:
- Remote collection of data needed for service and support activities
- Proactive system health status check
- Automatic reporting of Quality Club results to Phadia
Specifications
Specifications
| Type | Phadia 2500 Immunoassay Analyzer |
| Throughput | 3840 results per 8 hour shift |
| Ambient Temperature | 18°C to 32°C |
| Depth (English) | 48 in. |
| Height (English) | 77 in. |
| Width (English) | 163 in. |
| Voltage | 208 V |
| Mainframe Connection Protocol | ASTM, MasterCAP™, Phamas™ |
| Max. Networked Workstations | 5 systems of each type |
| Operating System | Windows™2000/ Windows XP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.