Learn More
Thermo Scientific™ Phadia™ EliA CCP Wells for Rheumatoid Arthritis
Catalog No. 14551501P Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 x 12 Tests
Catalog No. Quantity
14-551-501P 4 x 12 Tests
1 options
This item is not returnable. View return policy
Catalog No. 14-551-501P Supplier Thermo Scientific™ Supplier No. 14551501
Add to Cart
Add to Cart
This item is not returnable. View return policy
If present in the patient's specimen, IgG antibodies to CCP bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG antibodies are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA CCP Wells are coated with citrullinated synthetic peptides (second generation antigen)
  • The higher the response value, the more specific IgG is present in the specimen
  • EliA CCP IgA works with the same principle, but uses IgA conjugate
TRUSTED_SUSTAINABILITY
Specifications
Type EliA™ CCP Well
For Use With (Application) Rheumatoid Arthritis Assay
Quantity 4 x 12 Tests