Learn More
Thermo Scientific™ Phadia EliA™ CENP Wells for Connective Tissue Diseases
Catalog No. 14550501P Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 12 Tests
Catalog No. Quantity
14-550-501P 2 x 12 Tests
1 options
This item is not returnable. View return policy
Catalog No. 14-550-501P Supplier Thermo Scientific™ Supplier No. 14550501
Add to Cart
Add to Cart
This item is not returnable. View return policy
Intended for the in vitro quantitative measurement of IgG antibodies directed to CENP in human serum and plasma as an aid in the clinical diagnosis of scleroderma (CREST Syndrome).
If present in the patient's specimen, antibodies to CENP bind to their specific antigen.After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG antibodies (EliA IgG Conjugate) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA CENP Wells are coated with human recombinant centromere protein B
  • The higher the response value, the more specific IgG is present in the specimen
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Type EliA™ CENP Well
For Use With (Application) In-vitro quantitative measurement of IgG Antibodies directed to CENP in Human Serum and Plasma as an aid in the clinical diagnosis of scleroderma
Quantity 2 x 12 Tests