Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phadia™ EliA™ RF Wells for Rheumatoid Arthritis

Catalog No. 14560101P Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
EliA RF IgA Well
EliA RF IgM Well
Quantity:
4 x 12 Tests; Eq: 14-20 IU/mL
4 x 12 Tests; Eq: 3.5-5 IU/mL
For Use With (Application):
Measure IgA rheumatoid factor
Measure IgM rheumatoid factor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity For Use With (Application)
14-560-101P EliA RF IgA Well 4 x 12 Tests; Eq: 14-20 IU/mL Measure IgA rheumatoid factor
14-560-001P EliA RF IgM Well 4 x 12 Tests; Eq: 3.5-5 IU/mL Measure IgM rheumatoid factor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-560-101P Supplier Thermo Scientific™ Supplier No. 14560101

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Intended for the in vitro quantitative measurement of rheumatoid factor in serum and plasma to aid in the diagnosis of rheumatoid arthritis.

If present in the patient's specimen, rheumatoid factor binds to the antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgM (IgA) antibodies (EliA IgM, IgA Conjugate™) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.

  • The EliA RF IgM (IgA) Wells are coated with aggregated rabbit IgG (IgA) antigen
  • The higher the response value, the more specific IgM (IgA) is present in the specimen
  • All conjugates are isotype-specific -- mixed conjugate is not available

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Type EliA RF IgA Well
For Use With (Application) Measure IgA rheumatoid factor
Quantity 4 x 12 Tests; Eq: 14-20 IU/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.