Learn More
Thermo Scientific™ Phadia™ Gliadin Antibodies forCeliac, Inflammatory Bowel and Other Gastrointestinal Diseases
Catalog No. 14553901P Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
EliA GliadinDP IgA
EliA GliadinDP IgG
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-553-901P EliA GliadinDP IgG
14-553-801P EliA GliadinDP IgA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-553-901P Supplier Thermo Scientific™ Supplier No. 14553901
Add to Cart
Add to Cart
This item is not returnable. View return policy
Intended for the in vitro quantitative measurement of antibodies directed to gliadin and deamidated gliadin peptides, respectively, in serum or plasma to aid in the diagnosis of celiac disease
If present in the patient's specimen, antibodies to gliadin (gliadin peptides) bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG or IgA antibodies, respectively (EliA IgG, IgA Conjugate™), are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • EliA Gliadin IgG and EliA Gliadin IgA Wells are coated with purified gliadin antigen
  • EliA GliadinDP IgG and EliA GliadinDP IgA Wells are coated with synthetic deamidated gliadin peptides
  • The higher the response value, the more specific IgG or IgA, respectively, is present in the specimen
  • All conjugates are isotype-specific -- mixed conjugate is not available
TRUSTED_SUSTAINABILITY
Specifications
Type EliA GliadinDP IgG